Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153137.4 | NP_694777.3 | 15 | 1824 | ||
GenBank mRNA | AK031400.1 | - | 15 | 1764 | ||
AY383616.1 | - | 13 | 1408 | |||
BC137605.1 | - | 13 | 1435 | 8860624 | ||
BC171933.1 | - | 13 | 1435 | 9054217 |