Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153137.4 | NP_694777.3 | 12, 13 | 1606 | ||
GenBank mRNA | AK031400.1 | - | 12, 13 | 1546 | ||
AK161211.1 | - | 9, 10 | 1216 | |||
AY383616.1 | - | 10, 11 | 1190 | |||
BC036561.1 | - | 12, 13 | 1414 | |||
BC137605.1 | - | 10, 11 | 1217 | 8860624 | ||
BC171933.1 | - | 10, 11 | 1217 | 9054217 |