Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1f9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153511.3 | NP_705731.2 | 3 | 472 | ||
GenBank mRNA | AK081783.1 | - | 3 | 467 | ||
AK156687.1 | - | 469 | ||||
AY071843.1 | - | 2 | 198 | |||
BC116227.1 | - | 2 | 198 | 40091521 | ||
BC116228.1 | - | 2 | 198 | 40091522 |