Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrnp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153287.3 | NP_695019.1 | 5 | 1106 | ||
GenBank mRNA | AB091687.1 | - | 4 | 1078 | ||
AK029893.1 | - | 5 | 1373 | |||
AK140213.1 | - | 5 | 1119 | |||
AK171658.1 | - | 4 | 1171 | |||
BC029720.1 | - | 1 | 148 | |||
BC050066.1 | - | 5 | 1106 | 30023598 |