Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178676.4 | NP_848791.2 | 6 | 658 | ||
GenBank mRNA | AK032057.1 | - | 6 | 644 | ||
AK046218.1 | - | 6 | 634 | |||
AK087561.1 | - | 6 | 632 | |||
AK169947.1 | - | 1 | 68 | |||
AK170974.1 | - | 1 | 67 | |||
AY376710.1 | - | 6 | 626 | |||
AY714060.1 | - | 6 | 629 | |||
BC079871.1 | - | 6 | 629 | 6831925 |