Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam117a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172543.4 | NP_766131.2 | 8 | 1159 | ||
GenBank mRNA | AK077747.1 | - | 8 | 1160 | ||
AK143632.1 | - | 8 | 1167 | |||
AK167417.1 | - | 8 | 1160 | |||
BC055690.1 | - | 8 | 1131 | 6415669 |