Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002622.4 | NP_002613.2 | 2 | 128 | ||
XM_005268465.3 | XP_005268522.1 | 2 | 165 | |||
GenBank mRNA | AK223394.1 | - | 2 | 126 | ||
AK315353.1 | - | 2 | 114 | |||
BC003620.1 | - | 2 | 130 | 2959698 | ||
BC006202.1 | - | 2 | 130 | 2959725 | ||
BC011869.2 | - | 2 | 121 | 2959726 | ||
BG707314.1 | - | 2 | 142 | |||
Y17392.1 | - | 2 | 144 |