Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146004.1 | NP_666116.1 | 1 | 735 | ||
GenBank mRNA | BC024997.1 | - | 1 | 735 | 5008948 |