Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172546.2 | NP_766134.1 | 3 | 786 | ||
GenBank mRNA | AK033015.1 | - | 3 | 787 | ||
AK149951.1 | - | 2 | 85 | |||
AY151137.1 | - | 3 | 748 | |||
BC024086.1 | - | 2 | 134 |