Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009574.1 | NP_001009574.1 | 1 | 874 | ||
GenBank mRNA | BC139188.1 | - | 1 | 919 | 8862210 | |
BC139191.1 | - | 1 | 919 | 8862213 |
Set Membership: |
G-protein coupled receptor |