Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010830.1 | NP_001010830.1 | 1 | 59 |
Gene Symbol: Taar8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010837.1 | NP_001010837.1 | 1 | 59 | ||
GenBank mRNA | BC139105.1 | - | 1 | 85 | 8862127 | |
BC139106.1 | - | 1 | 85 | 8862128 |
Gene Symbol: Taar8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010840.2 | NP_001010840.1 | 1 | 98 | ||
GenBank mRNA | BC139121.1 | - | 1 | 98 | 8862143 | |
BC139122.1 | - | 1 | 98 | 8862144 |
Set Membership: |
G-protein coupled receptor |