Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfpl4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177783.1 | NP_001171254.1 | 2 | 400 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY726306.1 | - | 400 | Chr. - - [-] on Build |