Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002624.3 | NP_002615.2 | 4 | 390 | ||
NM_145897.2 | NP_665904.1 | 2 | 255 | |||
GenBank mRNA | AB055803.1 | - | 4 | 299 | ||
AB055804.1 | - | 3 | 267 | |||
AB055805.1 | - | 2 | 164 | |||
AK024094.1 | - | 3 | 1690 | |||
AK292623.1 | - | 4 | 299 | |||
BC003373.1 | - | 4 | 308 | |||
BC062671.1 | - | 4 | 302 | 6458093 | ||
BE392248.1 | - | 4 | 550 | |||
BT007195.1 | - | 4 | 273 |