Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC021785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001489.3 | NP_001001489.1 | 3 | 500 | ||
NM_001113385.1 | NP_001106856.1 | 3 | 500 | |||
NM_001113388.2 | NP_001106859.1 | 3 | 500 | |||
GenBank mRNA | AI527856.1 | - | 3 | 493 | ||
AK085266.1 | - | 3 | 501 | |||
AK165453.1 | - | 3 | 498 | |||
AK165493.1 | - | 3 | 497 | |||
BC021785.1 | - | 3 | 443 | |||
BC094014.1 | - | 2 | 227 | |||
BC147384.1 | - | 3 | 440 | |||
BC147385.1 | - | 3 | 440 |