Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172134.2 | NP_742146.1 | 2 | 220 | ||
GenBank mRNA | AK039194.1 | - | 2 | 202 | ||
AK080846.1 | - | 2 | 196 | |||
AK145470.1 | - | 2 | 184 | |||
AK166078.1 | - | 2 | 203 | |||
AK166464.1 | - | 2 | 185 | |||
BC027745.1 | - | 2 | 257 | 5064695 | ||
BC032171.1 | - | 2 | 191 | |||
BI143426.1 | - | 2 | 249 |