Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024539.1 | NP_001019710.1 | 12 | 1709 | ||
GenBank mRNA | AK032878.1 | - | 13 | 1394 | ||
BC059866.1 | - | 1 | 1585 |