Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163276.1 | NP_001156748.1 | 7 | 1157 | ||
NM_198107.2 | NP_932775.2 | 7 | 1160 | |||
GenBank mRNA | AK042340.1 | - | 7 | 1122 | ||
AK149345.1 | - | 7 | 1141 | |||
BC057056.1 | - | 7 | 1114 | 6813010 |