Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175450.4 | NP_780659.2 | 6, 7 | 825 | ||
GenBank mRNA | AK050106.1 | - | 6, 7 | 823 | ||
AK082695.1 | - | 6, 7 | 828 | |||
AK133716.1 | - | 6, 7 | 826 | |||
AK160713.1 | - | 6, 7 | 823 | |||
AK166113.1 | - | 823 | ||||
AK168691.1 | - | 6, 7 | 823 | |||
BC032968.1 | - | 5, 6 | 517 | |||
BC095984.1 | - | 6, 7 | 825 | 4978914 |