Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175450.4 | NP_780659.2 | 2 | 233 | ||
GenBank mRNA | AK032251.1 | - | 2 | 231 | ||
AK050106.1 | - | 2 | 231 | |||
AK082695.1 | - | 2 | 235 | |||
AK087505.1 | - | 1 | 864 | |||
AK133716.1 | - | 2 | 234 | |||
AK160713.1 | - | 2 | 231 | |||
AK163465.1 | - | 1 | 861 | |||
AK166113.1 | - | 231 | ||||
AK168691.1 | - | 2 | 231 | |||
BC095984.1 | - | 2 | 233 | 4978914 |