Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183152.3 | NP_898975.2 | 15 | 1857 | ||
GenBank mRNA | AK031968.1 | - | 15 | 1858 | ||
BC086684.1 | - | 10 | 1154 | 30679265 | ||
BC099679.1 | - | 15 | 1826 | 6489647 | ||
BU505736.1 | - | 4 | 466 |