Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153543.2 | NP_705771.2 | 7 | 969 | ||
GenBank mRNA | AK052361.1 | - | 7 | 860 | ||
AK086413.1 | - | 7 | 912 | |||
AK141954.1 | - | 7 | 931 | |||
AK215056.1 | - | 4 | 282 | |||
BC034531.1 | - | 7 | 891 | 4188055 |