Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153543.2 | NP_705771.2 | 19 | 2357 | ||
GenBank mRNA | AK052361.1 | - | 19 | 2248 | ||
AK141954.1 | - | 19 | 2318 | |||
AK144594.1 | - | 1 | 18 | |||
BC034531.1 | - | 19 | 2279 | 4188055 |