Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145220.2 | NP_660255.1 | 9 | 737 | ||
GenBank mRNA | AK154525.1 | - | 9 | 761 | ||
AK160075.1 | - | 9 | 716 | |||
AK160105.1 | - | 9 | 713 | |||
AK161003.1 | - | 9 | 744 | |||
AK168748.1 | - | 9 | 693 | |||
AK171512.1 | - | 9 | 693 | |||
AY113706.1 | - | 9 | 739 | |||
BC048906.1 | - | 9 | 675 | 6475576 |