Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018053.1 | NP_001018063.1 | 3 | 230 | ||
NM_006212.2 | NP_006203.2 | 3 | 230 | |||
GenBank mRNA | AB044805.1 | - | 3 | 183 | ||
AF470623.1 | - | 3 | 202 | |||
AJ005578.1 | - | 3 | 207 | |||
AK292883.1 | - | 3 | 255 | |||
AK302315.1 | - | 5 | 393 | |||
BC069350.1 | - | 3 | 171 | 7262370 | ||
BC069385.1 | - | 3 | 171 | 7262287 | ||
BC069583.1 | - | 3 | 171 | 7262299 | ||
BC069586.1 | - | 3 | 171 | 7262311 | ||
BC069600.1 | - | 3 | 171 | 7262323 | ||
BC075075.2 | - | 3 | 168 | 30915381 | ||
BC075076.2 | - | 3 | 168 | 30915594 | ||
BC112103.1 | - | 3 | 173 | 8327571 | ||
BC112105.1 | - | 3 | 173 | 8327573 | ||
BX510096.1 | - | 4 | 316 | |||
EU334168.1 | - | 3 | 248 |