Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207522.2 | NP_997405.2 | 3 | 555 | ||
GenBank mRNA | AK144620.1 | - | 3 | 547 | ||
AK148535.1 | - | 4 | 732 | |||
BC067068.1 | - | 3 | 529 | 30606425 |