Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024454.1 | NP_077774.1 | 3 | 528 | ||
GenBank mRNA | AB232615.1 | - | 3 | 283 | ||
AK035980.1 | - | 2 | 97 | |||
AK172891.1 | - | 3 | 528 | |||
BC055042.1 | - | 3 | 528 | 6490069 |