Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003950.2 | NP_001003950.1 | 5, 6 | 949 | ||
GenBank mRNA | AK152122.1 | - | 6, 7 | 1096 | ||
AK182637.1 | - | 1, 2 | 127 | |||
AK206753.1 | - | 1, 2 | 169 | |||
BC080289.1 | - | 5, 6 | 949 | 6816077 | ||
S79539.1 | - | 3, 4 | 1029 |