Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153059.1 | NP_694699.1 | 4 | 768 | ||
GenBank mRNA | AK043098.1 | - | 802 | |||
AK075928.1 | - | 4 | 758 | |||
AK078061.1 | - | 4 | 836 | |||
BC020100.1 | - | 4 | 768 | 3981114 | ||
BC023194.1 | - | 1 | 216 | |||
BC083125.1 | - | 5 | 936 |