Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115.2 | NP_001106.1 | 14, 15 | 3180 | ||
XM_005250769.3 | XP_005250826.1 | 13, 14 | 3213 | |||
XM_006716501.3 | XP_006716564.1 | 13, 14 | 3309 | |||
XM_017013006.1 | XP_016868495.1 | 12, 13 | 2981 | |||
GenBank mRNA | DQ104739.1 | - | 11, 12 | 2531 | ||
GQ472223.1 | - | 14, 15 | 5018 | |||
M83533.1 | - | 8, 9 | 1196 | |||
Z35309.1 | - | 14, 15 | 5018 |