Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003913.2 | NP_001003913.1 | 6 | 722 | ||
NM_001171582.1 | NP_001165053.1 | 6 | 722 | |||
GenBank mRNA | AK178137.1 | - | 4 | 362 | ||
AK184017.1 | - | 3 | 204 | |||
AK193348.1 | - | 3 | 234 | |||
AK214150.1 | - | 4 | 319 | |||
BC052042.1 | - | 6 | 574 | |||
BC058968.1 | - | 6 | 614 | |||
BC079643.1 | - | 6 | 575 | 6416029 |