Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033264.3 | NP_001028436.2 | 7, 8 | 936 | ||
NM_001285777.1 | NP_001272706.1 | 6, 7 | 856 | |||
NM_001285779.1 | NP_001272708.1 | 7, 8 | 965 | |||
GenBank mRNA | AK039618.1 | - | 6, 7 | 744 | ||
AK165896.1 | - | 7, 8 | 882 | |||
AK220231.1 | - | 7, 8 | 888 | |||
BC141115.1 | - | 7, 8 | 936 | 9055727 | ||
EU770627.1 | - | 7, 8 | 874 |