Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc85a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166661.2 | NP_001160133.1 | 2 | 1425 | ||
NM_181577.5 | NP_853555.2 | 2 | 1425 | |||
GenBank mRNA | AK045568.1 | - | 2 | 1398 | ||
AK085366.1 | - | 1 | 199 | |||
AK087049.1 | - | 2 | 1368 | |||
AK087070.1 | - | 2 | 1368 | |||
AK173298.1 | - | 2 | 1442 | |||
BC020949.1 | - | 1 | 388 | |||
BC066065.1 | - | 3 | 864 | 6850852 |