Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146015.2 | NP_666127.2 | 8 | 1022 | ||
GenBank mRNA | AK077302.1 | - | 8 | 1024 | ||
AY185605.1 | - | 7 | 847 | |||
AY251056.1 | - | 8 | 1017 | |||
BC031184.1 | - | 5 | 643 | 4989603 |