Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931440F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176829.2 | NP_789799.1 | 1 | 1903 | ||
GenBank mRNA | AK029918.1 | - | 1 | 1903 | ||
BC139107.1 | - | 1 | 1760 | 8862129 | ||
BC139123.1 | - | 1 | 1760 | 8862145 |
Gene Symbol: Fem1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010192.4 | NP_034322.3 | 1 | 1806 | ||
GenBank mRNA | AK005041.1 | - | 1 | 1765 | ||
AK009354.1 | - | 1 | 99 | |||
AK034412.1 | - | 1 | 1781 | |||
AK080978.1 | - | 1 | 1778 | |||
AK129000.1 | - | 1 | 1789 | |||
AK147879.1 | - | 1 | 1791 | |||
AK155363.1 | - | 1 | 1790 | |||
BC009161.1 | - | 1 | 1773 | 2811079 | ||
BC054382.1 | - | 1 | 1777 | 6469158 | ||
BC110669.1 | - | 1 | 1760 | 5042239 |