Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbq1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175000.2 | NP_778165.1 | 2 | 187 | ||
GenBank mRNA | AK039049.1 | - | 2 | 188 | ||
BC139349.1 | - | 2 | 161 | 8862371 | ||
BC139351.1 | - | 2 | 161 | 8862373 |
Gene Symbol: Hbq1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033981.3 | NP_001029153.1 | 2 | 187 | ||
GenBank mRNA | AK157060.1 | - | 2 | 188 | ||
BC141224.1 | - | 2 | 165 | 9055836 |