Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277305.1 | NP_001264234.1 | 9 | 1554 | ||
NM_175643.3 | NP_783574.1 | 9 | 1554 | |||
GenBank mRNA | AK040370.1 | - | 3 | 298 | ||
AK220464.1 | - | 8 | 1296 | |||
BC046456.1 | - | 9 | 1554 | 3965027 |