Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033597.1 | NP_001028769.1 | 21 | 2176 | ||
NM_001033598.1 | NP_001028770.1 | 21 | 2145 | |||
NM_001033599.1 | NP_001028771.1 | 21 | 2351 | |||
NM_144823.4 | NP_659072.3 | 21 | 2176 | |||
GenBank mRNA | AB073974.1 | - | 20 | 2020 | ||
AK122384.1 | - | 10 | 3942 | |||
AK147283.1 | - | 21 | 2146 | |||
AK147421.1 | - | 21 | 2177 | |||
AY167035.1 | - | 21 | 2146 | |||
AY786360.1 | - | 23 | 2694 | |||
AY786361.1 | - | 21 | 2126 | |||
AY786362.1 | - | 21 | 2318 | |||
AY786363.1 | - | 21 | 2171 | |||
BC016114.1 | - | 21 | 2102 | 4481949 | ||
BC022959.1 | - | 21 | 2176 | 5362660 |