Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033597.1 | NP_001028769.1 | 3 | 400 | ||
NM_001033598.1 | NP_001028770.1 | 3 | 369 | |||
NM_001033599.1 | NP_001028771.1 | 3 | 575 | |||
NM_144823.4 | NP_659072.3 | 3 | 400 | |||
GenBank mRNA | AB073974.1 | - | 2 | 244 | ||
AK032479.1 | - | 4 | 659 | |||
AK039380.1 | - | 3 | 486 | |||
AK147283.1 | - | 3 | 370 | |||
AK147421.1 | - | 3 | 401 | |||
AY167035.1 | - | 3 | 370 | |||
AY786360.1 | - | 5 | 918 | |||
AY786361.1 | - | 3 | 350 | |||
AY786362.1 | - | 3 | 542 | |||
AY786363.1 | - | 3 | 395 | |||
BB634066.1 | - | 3 | 575 | |||
BC016114.1 | - | 3 | 326 | 4481949 | ||
BC022959.1 | - | 3 | 400 | 5362660 |