Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033597.1 | NP_001028769.1 | 7 | 751 | ||
NM_001033598.1 | NP_001028770.1 | 7 | 720 | |||
NM_001033599.1 | NP_001028771.1 | 7 | 926 | |||
NM_144823.4 | NP_659072.3 | 7 | 751 | |||
GenBank mRNA | AB073974.1 | - | 6 | 595 | ||
AK032479.1 | - | 8 | 1010 | |||
AK039380.1 | - | 7 | 837 | |||
AK147283.1 | - | 7 | 721 | |||
AK147421.1 | - | 7 | 752 | |||
AY786360.1 | - | 9 | 1269 | |||
AY786361.1 | - | 7 | 701 | |||
AY786362.1 | - | 7 | 893 | |||
AY786363.1 | - | 7 | 746 | |||
BC016114.1 | - | 7 | 677 | 4481949 | ||
BC022959.1 | - | 7 | 751 | 5362660 |