Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iba57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173785.6 | NP_776146.1 | 3 | 1026 | ||
GenBank mRNA | AK038625.1 | - | 3 | 1021 | ||
AK133203.1 | - | 3 | 1024 | |||
AK172525.1 | - | 3 | 987 | |||
BC094909.1 | - | 3 | 1026 | 6432747 |