Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271356.1 | NP_001258285.1 | 4 | 768 | ||
NM_001271357.1 | NP_001258286.1 | 4 | 511 | |||
NM_146018.2 | NP_666130.1 | 4 | 808 | |||
GenBank mRNA | AK039106.1 | - | 4 | 595 | ||
AK080933.1 | - | 4 | 593 | |||
AK136071.1 | - | 4 | 499 | |||
AK148216.1 | - | 4 | 562 | |||
AK154040.1 | - | 4 | 790 | |||
AK161460.1 | - | 4 | 540 | |||
AK172245.1 | - | 4 | 567 | |||
BC025820.1 | - | 4 | 542 | 5099493 | ||
CJ052419.1 | - | 4 | 546 |