Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168507.1 | NP_001161979.1 | 3 | 968 | ||
NM_173453.3 | NP_775655.1 | 2 | 680 | |||
GenBank mRNA | AK077600.1 | - | 2 | 676 | ||
BC048514.1 | - | 2 | 428 | 6594842 | ||
CX211768.1 | - | 3 | 703 |