Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004143.4 | NP_001004143.2 | 4 | 683 | ||
GenBank mRNA | AK034492.1 | - | 4 | 658 | ||
AK040187.1 | - | 4 | 654 | |||
AK155071.1 | - | 4 | 684 | |||
AK160992.1 | - | 4 | 592 | |||
AK220368.1 | - | 4 | 644 | |||
BC080737.1 | - | 4 | 554 | 30431735 |