Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242339.1 | NP_001229268.1 | 22 | 2295 | ||
NM_001323067.1 | NP_001309996.1 | 20 | 2034 | |||
NM_001323068.1 | NP_001309997.1 | 19 | 2018 | |||
NM_001323069.1 | NP_001309998.1 | 20 | 2166 | |||
NM_001323070.1 | NP_001309999.1 | 14 | 1828 | |||
NM_001323071.1 | NP_001310000.1 | 21 | 2287 | |||
NM_001323072.1 | NP_001310001.1 | 21 | 2394 | |||
NM_001323073.1 | NP_001310002.1 | 16 | 1655 | |||
NM_001323074.1 | NP_001310003.1 | 15 | 1981 | |||
NM_002627.4 | NP_002618.1 | 20 | 2171 | |||
XM_005252465.4 | XP_005252522.1 | 21 | 2369 | |||
XM_005252466.4 | XP_005252523.1 | 20 | 2216 | |||
XM_005252467.2 | XP_005252524.1 | 20 | 2140 | |||
XM_006717449.1 | XP_006717512.1 | 21 | 2293 | |||
GenBank mRNA | AK092597.1 | - | 22 | 2295 | ||
AK097692.1 | - | 4 | 1224 | |||
AK128373.1 | - | 7 | 2683 | |||
AK129843.1 | - | 2027 | ||||
AK130767.1 | - | 5 | 1457 | |||
AK291841.1 | - | 20 | 2143 | |||
AK308003.1 | - | 20 | 2134 | |||
AK309692.1 | - | 13 | 1471 | |||
AL521453.3 | - | 4 | 346 | |||
BC002536.1 | - | 20 | 2131 | 3140233 | ||
BC021698.1 | - | 11 | 1084 | |||
BC029138.1 | - | 20 | 2171 | 5180268 | ||
BC043156.1 | - | 4 | 1867 | |||
D25328.1 | - | 20 | 2128 |