Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172560.3 | NP_766148.2 | 8 | 1989 | ||
GenBank mRNA | AK036710.1 | - | 7 | 1809 | ||
AK147774.1 | - | 8 | 1875 | |||
BC058512.1 | - | 8 | 822 | 6817014 |