Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172560.3 | NP_766148.2 | 19 | 3489 | ||
GenBank mRNA | AK036710.1 | - | 18 | 3309 | ||
AK147774.1 | - | 19 | 3379 | |||
BC058512.1 | - | 19 | 2322 | 6817014 |