Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145137.2 | NP_660119.1 | 2 | 221 | ||
GenBank mRNA | AK085751.1 | - | 1 | 81 | ||
AY103461.1 | - | 2 | 158 | |||
BC106833.1 | - | 1 | 147 | 40044847 | ||
BC110475.2 | - | 1 | 147 | 40045765 | ||
BC111520.1 | - | 1 | 147 | 40042306 | ||
BY198732.1 | - | 2 | 223 | |||
EU010011.1 | - | 2 | 153 |