Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005022.3 | NP_005013.1 | 3 | 1062 | ||
XM_017024761.1 | XP_016880250.1 | 2 | 1694 | |||
GenBank mRNA | AK312168.1 | - | 3 | 508 | ||
BC002475.2 | - | 3 | 492 | 3349355 | ||
BC006768.1 | - | 3 | 485 | 2900010 | ||
BC013439.1 | - | 3 | 485 | 3896948 | ||
BC015164.1 | - | 3 | 500 | 3890384 | ||
BC057828.1 | - | 3 | 511 | 30337970 | ||
BT007001.1 | - | 3 | 370 | |||
CR407670.1 | - | 3 | 370 | |||
GU727630.1 | - | 3 | 370 | |||
J03191.1 | - | 3 | 497 |