Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167669.1 | NP_001161141.1 | 2 | 233 | ||
NM_172561.3 | NP_766149.2 | 2 | 233 | |||
GenBank mRNA | AK042294.1 | - | 2 | 112 | ||
AK077543.1 | - | 2 | 111 | |||
AK166184.1 | - | 2 | 112 | |||
BC046751.1 | - | 2 | 94 | |||
BC055821.1 | - | 2 | 100 | 4502461 | ||
BY261455.1 | - | 2 | 108 |