Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139128.1 | NP_624354.1 | 8 | 1029 | ||
NM_139129.1 | NP_624355.1 | 7 | 909 | |||
NM_139130.2 | NP_624356.1 | 8 | 1029 | |||
GenBank mRNA | AB070894.1 | - | 8 | 1029 | ||
AB070895.1 | - | 8 | 1029 | |||
AB070896.1 | - | 7 | 909 | |||
AB288387.1 | - | 3 | 243 | |||
BC141247.1 | - | 7 | 986 | 9055859 | ||
BC145575.1 | - | 8 | 1107 |